Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005676.hg.1 |
ENST00000327443.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005676.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005676.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005676.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005676.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005676.hg.1 |
TC12000437.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000548560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000548560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000548560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000548560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000548560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000551969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000551969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000551969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
ENST00000551969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155243.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155243.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155243.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155243.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155243.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155243.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155243.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155243.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155243.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155243.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155243.hg.1 |
TC1200007713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200155242.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155242.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155242.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155242.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155242.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155242.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155242.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155242.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155242.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155242.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155242.hg.1 |
TC1200007713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000327443.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000426431.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000548560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000548560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000548560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000548560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000548560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000551969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000551969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000551969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
ENST00000551969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_001251825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_003109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007713.hg.1 |
NM_138473.2 |
Refseq |