Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020120.3 | NP_064505.1 | 33 - 34 | 3890 | 79 | |
NR_027671.2 | 33 - 34 | 4043 | 79 | |||
XM_006712634.3 | XP_006712697.1 | 33 - 34 | 3864 | 79 | ||
XM_006712635.3 | XP_006712698.1 | 32 - 33 | 3834 | 79 | ||
XM_006712636.3 | XP_006712699.1 | 33 - 34 | 3853 | 79 | ||
XM_017004507.1 | XP_016859996.1 | 34 - 35 | 3949 | 79 | ||
XM_017004508.1 | XP_016859997.1 | 34 - 35 | 3904 | 79 | ||
XM_017004509.1 | XP_016859998.1 | 19 - 20 | 2195 | 79 | ||
XM_017004510.1 | XP_016859999.1 | 15 - 16 | 2574 | 79 | ||
GenBank mRNA | AF227905.1 | - | 33 - 34 | 3750 | 79 | |
AK023671.1 | - | 14 - 15 | 1586 | 79 | ||
AK074251.1 | - | 17 - 18 | 1914 | 79 | ||
AK074376.1 | - | 7 - 8 | 885 | 79 | ||
AK293066.1 | - | 33 - 34 | 4043 | 79 | ||
BC041098.1 | - | 33 - 34 | 4040 | 5492334 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |