Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019440.hg.1 |
TC05001392.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019440.hg.1 |
ENST00000265036.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019440.hg.1 |
ENST00000453022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019440.hg.1 |
ENST00000545085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019440.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019440.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007533.hg.1 |
ENST00000502277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000505017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000505017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000505017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000509006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000509006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175734.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175734.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175734.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175734.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175734.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175734.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175734.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175734.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175734.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175734.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175734.hg.1 |
TC0500010865.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000265036.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000453022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000505017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000505017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000505017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000509006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000509006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
ENST00000512452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_001145208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010865.hg.1 |
NM_018369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007533.hg.1 |
ENST00000502277.1 |
Ensembl |