Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013940.hg.1 |
TC02000954.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013940.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013940.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000461704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000461704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000463418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
ENST00000463418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000461704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000461704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000463418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
ENST00000463418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009739.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167984.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167984.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167984.hg.1 |
ENST00000259056.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167984.hg.1 |
NM_014568.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167984.hg.1 |
TC0200009739.hg.1 |
Clariom D TC |