Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010402.hg.1 |
TC05000726.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010402.hg.1 |
ENST00000235510.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010402.hg.1 |
ENST00000253810.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010402.hg.1 |
ENST00000297183.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010402.hg.1 |
ENST00000310331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010402.hg.1 |
ENST00000310356.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010402.hg.1 |
ENST00000422011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010402.hg.1 |
ENST00000532219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010402.hg.1 |
NM_003732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010402.hg.1 |
NM_020690.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
ENST00000310331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
ENST00000437495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
ENST00000474060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
ENST00000532219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
NM_003732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
NM_020690.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
ENST00000310331.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
ENST00000437495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
ENST00000474060.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
ENST00000532219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
NM_003732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
NM_020690.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161298.hg.1 |
TC0500008854.hg.1 |
Clariom D TC |