Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016231.4 | NP_057315.3 | 3 - 4 | 860 | 100 | |
XM_005257988.2 | XP_005258045.1 | 3 - 4 | 875 | 100 | ||
GenBank mRNA | AF180819.1 | - | 3 - 4 | 858 | 100 | |
AF197898.1 | - | 3 - 4 | 860 | 100 | ||
AK315315.1 | - | 3 - 4 | 875 | 100 | ||
BC064663.1 | - | 3 - 4 | 1372 | 6527673 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |