Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008198.hg.1 |
ENST00000582820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008198.hg.1 |
ENST00000582820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003594.hg.1 |
ENST00000267843.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003594.hg.1 |
ENST00000560704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003594.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003594.hg.1 |
TC15000386.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560704.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
ENST00000560704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
ENST00000560704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151227.hg.1 |
TC1500007185.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560704.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007185.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
ENST00000267843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
ENST00000560270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
ENST00000560765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
ENST00000560979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
ENST00000614567.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
NM_002009.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151223.hg.1 |
TC1500007185.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1800008198.hg.1 |
ENST00000582820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008198.hg.1 |
ENST00000582820.1 |
Ensembl |