Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000471400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000471400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000471400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
ENST00000471400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000471400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000471400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000471400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
ENST00000471400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007817.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
ENST00000288065.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
ENST00000373866.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
ENST00000373869.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
NM_001286574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
NM_001286576.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
NM_145028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154126.hg.1 |
TC0600007817.hg.1 |
Clariom D TC |