Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000460311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000460311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
ENST00000460311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037958.hg.1 |
TC02002415.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037958.hg.1 |
ENST00000303319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037958.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037958.hg.1 |
ENST00000428879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037958.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200205962.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205962.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205962.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205962.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200205962.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205962.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205962.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205962.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200205962.hg.1 |
TC0200014584.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000303319.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000422782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000428879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000460311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000460311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
ENST00000460311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014584.hg.1 |
NM_015702.2 |
Refseq |