Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
ENST00000398215.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006917.hg.1 |
NM_000496.2 |
Refseq |