Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019090.hg.1 |
ENST00000333461.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019090.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019090.hg.1 |
TC17001447.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183252.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183252.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183252.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183252.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183252.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183252.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183252.hg.1 |
TC1700010564.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |