Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034828.hg.1 |
TC03001948.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034828.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034828.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000482640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000482640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000482640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
ENST00000482640.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196717.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196717.hg.1 |
ENST00000482640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196717.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196717.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196717.hg.1 |
ENST00000482640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196717.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196717.hg.1 |
TC0300012943.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000264265.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000482640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000482640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000482640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
ENST00000482640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012943.hg.1 |
NM_020169.3 |
Refseq |