Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000498253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000498253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
ENST00000498253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054965.hg.1 |
TC01003544.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054965.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054965.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054965.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054970.hg.1 |
TC01003544.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054970.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054970.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054967.hg.1 |
TC01003544.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054967.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054967.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054967.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225226.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225226.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225226.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225226.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225226.hg.1 |
TC0100016470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000498253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000498253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
ENST00000498253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016470.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225220.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225220.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225220.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225220.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225220.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225220.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225220.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225220.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225220.hg.1 |
TC0100016470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100225222.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225222.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225222.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225222.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225222.hg.1 |
ENST00000367677.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225222.hg.1 |
ENST00000476371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225222.hg.1 |
NM_022100.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225222.hg.1 |
NR_037606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225222.hg.1 |
TC0100016470.hg.1 |
Clariom D TC |