Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013081.hg.1 |
TC01000822.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013081.hg.1 |
ENST00000234701.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013081.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013081.hg.1 |
ENST00000495894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013081.hg.1 |
ENST00000539889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013081.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163643.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163643.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163643.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163643.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163643.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163643.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163643.hg.1 |
TC0100008922.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000234701.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
ENST00000394711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008922.hg.1 |
NM_001285.3 |
Refseq |