Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278298.1 | NP_001265227.1 | 27 - 28 | 5533 | 97 | |
NM_153264.6 | NP_694996.5 | 27 - 28 | 5533 | 97 | ||
NR_022012.2 | 27 - 28 | 5533 | 97 | |||
XM_011512621.2 | XP_011510923.1 | 26 - 27 | 5344 | 97 | ||
XM_011512622.2 | XP_011510924.1 | 27 - 28 | 5407 | 97 | ||
XM_011512623.2 | XP_011510925.1 | 27 - 28 | 5407 | 97 | ||
GenBank mRNA | AM906081.1 | - | 19 - 20 | 1475 | 97 | |
EU085556.1 | - | 27 - 28 | 5533 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |