Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026445.hg.1 |
ENST00000302438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026445.hg.1 |
ENST00000339824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026445.hg.1 |
ENST00000425217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026445.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026445.hg.1 |
TC12002015.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000543793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000543793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000543793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194100.hg.1 |
TC1200012100.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000339824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000425217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000543793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000543793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000543793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
ENST00000545002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012100.hg.1 |
NM_173598.4 |
Refseq |