Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL21A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318751.1 | NP_001305680.1 | 22 - 23 | 2327 | 109 | |
NM_001318752.1 | NP_001305681.1 | 20 - 21 | 2208 | 109 | ||
NM_001318753.1 | NP_001305682.1 | 5 - 6 | 374 | 109 | ||
NM_001318754.1 | NP_001305683.1 | 5 - 6 | 374 | 109 | ||
NM_030820.3 | NP_110447.2 | 21 - 22 | 2253 | 109 | ||
NR_134849.1 | 21 - 22 | 2245 | 109 | |||
NR_134850.1 | 22 - 23 | 2270 | 109 | |||
NR_134851.1 | 22 - 23 | 2278 | 109 | |||
XM_006715223.1 | XP_006715286.1 | 21 - 22 | 2419 | 109 | ||
XM_011514924.2 | XP_011513226.1 | 21 - 22 | 2196 | 109 | ||
XM_011514925.2 | XP_011513227.1 | 23 - 24 | 2366 | 109 | ||
XM_011514926.1 | XP_011513228.1 | 21 - 22 | 2124 | 109 | ||
XM_011514927.1 | XP_011513229.1 | 21 - 22 | 2078 | 109 | ||
GenBank mRNA | AF330693.1 | - | 20 - 21 | 2174 | 109 | |
AF370383.1 | - | 9 - 10 | 509 | 109 | ||
AF414088.1 | - | 21 - 22 | 109 | |||
AK096444.1 | - | 5 - 6 | 377 | 109 | ||
AK126586.1 | - | 22 - 23 | 2138 | 109 | ||
AK313398.1 | - | 21 - 22 | 2229 | 109 | ||
AL136624.1 | - | 21 - 22 | 109 | |||
BC045597.1 | - | 21 - 22 | 2200 | 4814199 | 109 | |
BC126108.1 | - | 21 - 22 | 2108 | 8991824 | 109 | |
BC143862.1 | - | 21 - 22 | 2100 | 109 | ||
BC143863.1 | - | 22 - 23 | 2125 | 109 | ||
BC143864.1 | - | 22 - 23 | 2133 | 109 | ||
BC143865.1 | - | 22 - 23 | 2182 | 9052381 | 109 | |
DA826579.1 | - | 5 - 6 | 364 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |