Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000463090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000463090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000463090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000463090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
ENST00000463090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000273.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000273.hg.1 |
ENST00000420960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000273.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000273.hg.1 |
TC20000023.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000144595.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144595.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144595.hg.1 |
TC2000006508.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144595.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144595.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000381458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000463090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000463090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000463090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000463090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
ENST00000463090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006508.hg.1 |
NM_003245.3 |
Refseq |