Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026807.hg.1 |
TC03001472.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026807.hg.1 |
ENST00000315251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026807.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026810.hg.1 |
TC03001472.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026810.hg.1 |
ENST00000315251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026810.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026811.hg.1 |
TC03001472.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026811.hg.1 |
ENST00000315251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026811.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026806.hg.1 |
TC03001472.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026806.hg.1 |
ENST00000315251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026806.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026804.hg.1 |
TC03001472.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026804.hg.1 |
ENST00000315251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026804.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026805.hg.1 |
TC03001472.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026805.hg.1 |
ENST00000315251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026805.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184026.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184026.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184026.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184026.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184026.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184026.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184026.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184026.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184026.hg.1 |
TC0300011220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300184024.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184024.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184024.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184024.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184024.hg.1 |
TC0300011220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300184032.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184032.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184032.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184032.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184032.hg.1 |
TC0300011220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300184025.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184025.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184025.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184025.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184025.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184025.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184025.hg.1 |
TC0300011220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300184028.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184028.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184028.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184028.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184028.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184028.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184028.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184028.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184028.hg.1 |
TC0300011220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011220.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184031.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184031.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184031.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184031.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184031.hg.1 |
TC0300011220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300184027.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184027.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184027.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184027.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184027.hg.1 |
ENST00000315251.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184027.hg.1 |
ENST00000467802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184027.hg.1 |
ENST00000481668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184027.hg.1 |
NM_018397.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184027.hg.1 |
TC0300011220.hg.1 |
Clariom D TC |