Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000469106.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000469106.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000469106.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000469106.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
ENST00000469106.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022664.hg.1 |
TC09001754.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022664.hg.1 |
ENST00000312665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022664.hg.1 |
ENST00000355097.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022664.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022664.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182737.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182737.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182737.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182737.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182737.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182737.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182737.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182737.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182737.hg.1 |
TC0900012029.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000312665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000355097.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000460614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000469106.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000469106.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000469106.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000469106.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
ENST00000469106.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_001246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012029.hg.1 |
NM_203468.2 |
Refseq |