Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
TC03000047.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
ENST00000295981.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
ENST00000383812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
ENST00000403601.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
ENST00000413608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
ENST00000416074.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
ENST00000455057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
ENST00000498214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
NM_001203263.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
NM_001203264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
NM_001203265.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
NM_032732.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
NM_153460.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
NM_153461.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001084.hg.1 |
NR_037807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001087.hg.1 |
TC03000047.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001087.hg.1 |
ENST00000498214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001087.hg.1 |
NR_037807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000295981.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000383812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000403601.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000412901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000413608.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000416074.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000424206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000434756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000436503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000438091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000451165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000451231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000451271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000455057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000466046.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000476810.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000478206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000483582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000490512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000494365.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000498214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_001203263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_001203264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_001203265.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_032732.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_153460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_153461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NR_037807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000295981.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000383812.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000403601.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000412901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000413608.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000416074.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000424206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000434756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000436503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000438091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000451165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000451231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000451271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000455057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000466046.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000476810.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000478206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000483582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000490512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000494365.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
ENST00000498214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_001203263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_001203264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_001203265.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_032732.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_153460.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NM_153461.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
NR_037807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145437.hg.1 |
TC0300006565.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000412901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000464406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000466046.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000478206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000490512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000494365.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000498214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
NR_037807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000412901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000464406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000466046.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000478206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000490512.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000494365.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
ENST00000498214.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
NR_037807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145440.hg.1 |
TC0300006565.hg.1 |
Clariom D TC |