Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014274.hg.1 |
ENST00000225728.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014274.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014274.hg.1 |
TC17001066.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700174104.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174104.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174104.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174104.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700174104.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174104.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174104.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174104.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700174104.hg.1 |
TC1700009592.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000225728.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000574128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
ENST00000575519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009592.hg.1 |
NM_016060.2 |
Refseq |