Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000490884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000490884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000490884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000490884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000498802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000498802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
ENST00000498802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013047.hg.1 |
TC01000821.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013047.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013047.hg.1 |
ENST00000439777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013047.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013049.hg.1 |
TC01000821.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013049.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013049.hg.1 |
ENST00000439777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013049.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163617.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163617.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163617.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163617.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163617.hg.1 |
TC0100008921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000370565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000490884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000490884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000490884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000490884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000498802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000498802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
ENST00000498802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008921.hg.1 |
NM_006536.6 |
Refseq |