Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005571.hg.1 |
TC03000276.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005571.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005571.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151891.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151891.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151891.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151891.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151891.hg.1 |
TC0300007324.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
ENST00000296435.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007324.hg.1 |
NM_004345.4 |
Refseq |