Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000548675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000548675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000548675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000548675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000550434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000550434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000550434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000550434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000550434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000551415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
ENST00000551415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004656.hg.1 |
ENST00000257981.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004656.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004656.hg.1 |
TC12000374.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000548675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000548675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000548675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000548675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000550434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000550434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000550434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000550434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000550434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000551415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
ENST00000551415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007567.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200153341.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153341.hg.1 |
ENST00000548675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153341.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200153341.hg.1 |
ENST00000257981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153341.hg.1 |
ENST00000548675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153341.hg.1 |
NM_012284.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200153341.hg.1 |
TC1200007567.hg.1 |
Clariom D TC |