Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033539.hg.1 |
TC01002178.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033539.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033539.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194533.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194533.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194533.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194533.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194533.hg.1 |
TC0100012786.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
ENST00000400906.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012786.hg.1 |
NM_207420.2 |
Refseq |