Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020427.hg.1 |
ENST00000370418.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020427.hg.1 |
ENST00000472355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020427.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020427.hg.1 |
TC10001586.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000179085.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179085.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179085.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179085.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179085.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179085.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179085.hg.1 |
TC1000011599.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000370418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
ENST00000441382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011599.hg.1 |
NM_001308.2 |
Refseq |