Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005336.hg.1 |
TC08000374.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005336.hg.1 |
ENST00000260102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005336.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005333.hg.1 |
TC08000374.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005333.hg.1 |
ENST00000260102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005333.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005332.hg.1 |
TC08000374.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005332.hg.1 |
ENST00000260102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005332.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005335.hg.1 |
TC08000374.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005335.hg.1 |
ENST00000260102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005335.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152652.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152652.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152652.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152652.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152652.hg.1 |
TC0800007653.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800152657.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152657.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152657.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152657.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152657.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152657.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152657.hg.1 |
TC0800007653.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007653.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152656.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152656.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152656.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152656.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152656.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152656.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152656.hg.1 |
ENST00000522521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152656.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152656.hg.1 |
TC0800007653.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800152654.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152654.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152654.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152654.hg.1 |
ENST00000260102.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152654.hg.1 |
ENST00000519831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152654.hg.1 |
NM_014175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152654.hg.1 |
TC0800007653.hg.1 |
Clariom D TC |