Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001271.hg.1 |
TC05000132.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001271.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001271.hg.1 |
ENST00000514738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001271.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001272.hg.1 |
TC05000132.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001272.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001272.hg.1 |
ENST00000514738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001272.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000504835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000504835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000504835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000504835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000504835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000506396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000506396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000506396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000506396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000508172.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147539.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147539.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147539.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147539.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147539.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147539.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147539.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147539.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147539.hg.1 |
TC0500007044.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000504835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000504835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000504835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000504835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000504835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000506396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000506396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000506396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000506396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000508172.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007044.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147540.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147540.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147540.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147540.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147540.hg.1 |
ENST00000265071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147540.hg.1 |
ENST00000508132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147540.hg.1 |
ENST00000514738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147540.hg.1 |
NM_004932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147540.hg.1 |
TC0500007044.hg.1 |
Clariom D TC |