Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010483.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010483.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171204.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171204.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171204.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171204.hg.1 |
ENST00000625168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171204.hg.1 |
NM_001039380.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171204.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171204.hg.1 |
TC1000010483.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000171202.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171202.hg.1 |
NR_126421.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171202.hg.1 |
TC1000010483.hg.1 |
Clariom D TC |