Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195446.1 | NP_001182375.1 | 5 - 6 | 808 | 123 | |
GenBank mRNA | AK297161.1 | - | 5 - 6 | 675 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |