Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000478135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000478135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000478135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000478135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000478135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000479232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000479232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000479232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000479232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000494750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000494750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000494750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
ENST00000494750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000255945.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000461940.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000478135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000478135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000478135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000478135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000478135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000479232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000479232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000479232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000479232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000494750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000494750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000494750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
ENST00000494750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009677.hg.1 |
NM_018326.2 |
Refseq |