Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013059.hg.1 |
TC0X000980.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013059.hg.1 |
ENST00000333807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013059.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164846.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164846.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164846.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164846.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164846.hg.1 |
TC0X00009493.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000343571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
ENST00000420999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009493.hg.1 |
NM_025184.3 |
Refseq |