Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015822.hg.1 |
ENST00000253115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015822.hg.1 |
ENST00000535489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015822.hg.1 |
ENST00000545189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015822.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015822.hg.1 |
TC19001148.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009586.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
NM_024106.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
TC1900009586.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
ENST00000253115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
ENST00000535489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
ENST00000589289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
ENST00000593003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
NM_001300883.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177174.hg.1 |
NM_024106.2 |
Refseq |