Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001177.hg.1 |
ENST00000380152.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001177.hg.1 |
ENST00000544455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001177.hg.1 |
NM_000059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001177.hg.1 |
TC13000111.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300146310.hg.1 |
ENST00000380152.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146310.hg.1 |
ENST00000544455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146310.hg.1 |
NM_000059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146310.hg.1 |
ENST00000380152.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146310.hg.1 |
ENST00000544455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146310.hg.1 |
NM_000059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146310.hg.1 |
TC1300006802.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300146309.hg.1 |
ENST00000380152.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146309.hg.1 |
ENST00000544455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146309.hg.1 |
NM_000059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146309.hg.1 |
ENST00000380152.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146309.hg.1 |
ENST00000544455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146309.hg.1 |
NM_000059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146309.hg.1 |
TC1300006802.hg.1 |
Clariom D TC |