Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026233.hg.1 |
ENST00000405440.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026233.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026233.hg.1 |
TC12001996.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026225.hg.1 |
ENST00000310346.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026225.hg.1 |
ENST00000349716.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026225.hg.1 |
ENST00000405440.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026225.hg.1 |
ENST00000448888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026225.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026225.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026225.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026225.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026225.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026225.hg.1 |
TC12001996.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026226.hg.1 |
ENST00000310346.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026226.hg.1 |
ENST00000349716.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026226.hg.1 |
ENST00000405440.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026226.hg.1 |
ENST00000448888.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026226.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026226.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026226.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026226.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026226.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026226.hg.1 |
TC12001996.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200193576.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193576.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193576.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193576.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193576.hg.1 |
TC1200012008.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200193577.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193577.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193577.hg.1 |
TC1200012008.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012008.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
ENST00000310346.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
ENST00000349716.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
ENST00000405440.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
ENST00000526441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
ENST00000552726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
NM_000192.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
NM_080717.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
NM_181486.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193565.hg.1 |
TC1200012008.hg.1 |
Clariom D TC |