Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05021610.hg.1 |
TC05001590.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05021610.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05021610.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000505631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000505631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000505631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000505631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000507075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000507075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000507075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000507075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000508948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000508948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000508948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000508948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000511391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000511391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000511391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000514284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000514284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000514284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
ENST00000514284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500179249.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500179249.hg.1 |
ENST00000511391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500179249.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500179249.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500179249.hg.1 |
ENST00000511391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500179249.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500179249.hg.1 |
TC0500011448.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000265138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000503192.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000505631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000505631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000505631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000505631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000507075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000507075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000507075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000507075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000508948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000508948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000508948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000508948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000511391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000511391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000511391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000514284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000514284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000514284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
ENST00000514284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011448.hg.1 |
NM_020801.3 |
Refseq |