Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011527471.2 | XP_011525773.1 | 17 - 18 | 2012 | 112 | |
XM_011527472.2 | XP_011525774.1 | 17 - 18 | 2012 | 112 | ||
XM_011527473.2 | XP_011525775.1 | 17 - 18 | 2012 | 112 | ||
XM_011527474.2 | XP_011525776.1 | 16 - 17 | 1892 | 112 | ||
XM_011527475.2 | XP_011525777.1 | 17 - 18 | 2012 | 112 | ||
XM_011527476.2 | XP_011525778.1 | 17 - 18 | 2012 | 112 | ||
XM_017027458.1 | XP_016882947.1 | 17 - 18 | 2012 | 112 | ||
XM_017027459.1 | XP_016882948.1 | 7 - 8 | 995 | 112 | ||
GenBank mRNA | BC049000.1 | - | 17 - 18 | 2252 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |