Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004189.3 | NP_004180.1 | 1 | 1142 | 98 | |
GenBank mRNA | AK314127.1 | - | 1 | 1130 | 98 | |
BC106729.1 | - | 1 | 691 | 40015392 | 98 | |
BC106730.1 | - | 1 | 691 | 40015393 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon |