Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015759.hg.1 |
TC06001320.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015759.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015759.hg.1 |
ENST00000438606.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015759.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015759.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015758.hg.1 |
TC06001320.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015758.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015758.hg.1 |
ENST00000438606.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015758.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015758.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174703.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174703.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174703.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174703.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174703.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174703.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174703.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174703.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174703.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174703.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174703.hg.1 |
TC0600011037.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600174704.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174704.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174704.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174704.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174704.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174704.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174704.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174704.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174704.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174704.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174704.hg.1 |
TC0600011037.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000306482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000615510.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
ENST00000617911.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_000948.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011037.hg.1 |
NM_001163558.2 |
Refseq |