Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001230.hg.1 |
ENST00000302926.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001230.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001230.hg.1 |
TC17000093.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000570940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000570940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000570940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000570940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000572893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000572893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000570940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000570940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000570940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000570940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000572893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000572893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006744.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146904.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146904.hg.1 |
ENST00000570940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146904.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146904.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146904.hg.1 |
ENST00000302926.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146904.hg.1 |
ENST00000570940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146904.hg.1 |
ENST00000575301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146904.hg.1 |
NM_020795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146904.hg.1 |
TC1700006744.hg.1 |
Clariom D TC |