Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AACS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK022740.1 | - | 12 - 13 | 1813 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |