Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000733.3 | NP_000724.1 | 4 - 5 | 342 | 120 | |
GenBank mRNA | AK292612.1 | - | 4 - 5 | 191 | 120 | |
AK301255.1 | - | 4 - 5 | 192 | 120 | ||
BC049847.1 | - | 3 - 4 | 154 | 5214563 | 120 | |
BI910359.1 | - | 4 - 5 | 377 | 120 | ||
BP366535.1 | - | 4 - 5 | 359 | 120 | ||
X03884.1 | - | 3 - 4 | 140 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |