Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018201.4 | NP_060671.3 | 9 - 10 | 1066 | 110 | |
XM_005252060.2 | XP_005252117.1 | 8 - 9 | 969 | 110 | ||
GenBank mRNA | AB209565.1 | - | 6 - 7 | 785 | 110 | |
AB449899.1 | - | 9 - 10 | 919 | 110 | ||
AK027457.1 | - | 1 - 2 | 110 | 110 | ||
AK123828.1 | - | 5 - 6 | 680 | 110 | ||
AK289777.1 | - | 9 - 10 | 978 | 110 | ||
AK295183.1 | - | 6 - 7 | 769 | 110 | ||
AK307556.1 | - | 5 - 6 | 668 | 110 | ||
BC014898.1 | - | 9 - 10 | 941 | 110 | ||
BC132817.1 | - | 9 - 10 | 939 | 40146839 | 110 | |
BC136509.1 | - | 9 - 10 | 939 | 9020499 | 110 | |
BC150311.1 | - | 9 - 10 | 975 | 8860447 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |