Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004084.hg.1 |
ENST00000321505.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004084.hg.1 |
ENST00000389726.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004084.hg.1 |
ENST00000536568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004084.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004084.hg.1 |
TC11000317.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100150955.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150955.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150955.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150955.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150955.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150955.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150955.hg.1 |
TC1100007233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000265654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000321505.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
ENST00000526400.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007233.hg.1 |
NM_012194.2 |
Refseq |