Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022481.5 | NP_071926.4 | 18 - 19 | 2715 | 56 | |
XM_005268497.1 | XP_005268554.1 | 18 - 19 | 2679 | 56 | ||
XM_005268498.1 | XP_005268555.1 | 18 - 19 | 2679 | 56 | ||
XM_005268499.1 | XP_005268556.1 | 18 - 19 | 2653 | 56 | ||
XM_005268500.3 | XP_005268557.1 | 18 - 19 | 2679 | 56 | ||
XM_006714792.1 | XP_006714855.1 | 18 - 19 | 2679 | 56 | ||
XM_011537677.2 | XP_011535979.1 | 16 - 17 | 2134 | 56 | ||
GenBank mRNA | AJ310567.1 | - | 17 - 18 | 2665 | 56 | |
AK001579.1 | - | 2 - 3 | 386 | 56 | ||
AK091385.1 | - | 18 - 19 | 2594 | 56 | ||
AK290052.1 | - | 18 - 19 | 2708 | 56 | ||
AK295768.1 | - | 12 - 13 | 1692 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Probe spans exons |