Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057429.hg.1 |
TC01003743.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057429.hg.1 |
ENST00000367161.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057429.hg.1 |
ENST00000367162.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057429.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057429.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017060.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229207.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229207.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229207.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229207.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229207.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229207.hg.1 |
ENST00000367161.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229207.hg.1 |
ENST00000367162.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229207.hg.1 |
ENST00000615388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229207.hg.1 |
NM_015375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229207.hg.1 |
NM_199462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229207.hg.1 |
TC0100017060.hg.1 |
Clariom D TC |