Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005268151.3 | XP_005268208.1 | 2 | 11622 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon |