Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052883.1 | NP_443115.1 | 13 - 14 | 1770 | 105 | |
GenBank mRNA | AF133519.1 | - | 13 - 14 | 1438 | 105 | |
AF171055.1 | - | 13 - 14 | 1444 | 105 | ||
BC030028.2 | - | 13 - 14 | 1748 | 105 | ||
BC050032.1 | - | 13 - 14 | 1757 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |