Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011329.hg.1 |
TC04000819.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011329.hg.1 |
ENST00000358572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011329.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011329.hg.1 |
ENST00000513272.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011329.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163170.hg.1 |
TC0400009179.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000358572.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000503148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000506214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000508652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000509657.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000511562.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
ENST00000513272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009179.hg.1 |
NM_018352.2 |
Refseq |